Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aurora A/B Monoclonal Antibody (3H1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547320
Description
Aurora A/B Monoclonal Antibody for Western Blot, ICC/IF, IHC
The Aurora gene, a serine-threonine kinase, orginally discovered in Drosophila, has been found to be responsible for a chromosomal segregation defect. The mammalian homologues of Drosophila Aurora have been termed ARK1 (Aurora-related kinase 1) and ARK2.
Specifications
Aurora A/B | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 5 mM sodium azide | |
O14965, O55099, O70126, P59241, P97477, Q96GD4 | |
Mouse | |
Protein A/G | |
RUO | |
114592, 20877, 20878, 261730, 6790, 9212 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
3H1 | |
Unconjugated | |
AURKA | |
AURKA, AURKB | |
Full length recombinant human Aurora A protein expressed in and purified from E. coli. | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction