Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aurora A/B Monoclonal Antibody (3H1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547418
Description
Aurora A/B Monoclonal Antibody for Western Blot, ICC/IF, IHC
The Aurora gene, a serine-threonine kinase, orginally discovered in Drosophila, has been found to be responsible for a chromosomal segregation defect. The mammalian homologues of Drosophila Aurora have been termed ARK1 (Aurora-related kinase 1) and ARK2.
Specifications
Aurora A/B | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 5 mM sodium azide | |
O14965, O55099, O70126, P59241, P97477, Q96GD4 | |
Mouse | |
Protein A/G | |
RUO | |
114592, 20877, 20878, 261730, 6790, 9212 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
3H1 | |
Unconjugated | |
AURKA | |
AURKA, AURKB | |
Full length recombinant human Aurora A protein expressed in and purified from E. coli. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction