Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aurora A/B Monoclonal Antibody (5A12)
GREENER_CHOICE

Catalog No. PIMA547439
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547439 100 μL
PIMA547340 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA547439 Supplier Invitrogen™ Supplier No. MA547439
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Aurora A/B Monoclonal Antibody for Western Blot, ICC/IF, IHC

The Aurora gene, a serine-threonine kinase, orginally discovered in Drosophila, has been found to be responsible for a chromosomal segregation defect. The mammalian homologues of Drosophila Aurora have been termed ARK1 (Aurora-related kinase 1) and ARK2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aurora A/B
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5A12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene AURKA
Gene Accession No. O14965, O55099, O70126, P59241, P97477, Q96GD4
Gene Symbols AURKA, AURKB
Host Species Mouse
Immunogen Full length recombinant human Aurora A protein expressed in and purified from E. coli.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050161, 100073050, 114592, 20877, 20878, 261730, 479492, 485940, 6790, 9212
Target Species Human, Mouse, Rat, Canine, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.