Learn More
Promega Aurora A Kinase Enzyme System, 10microg
Catalog No. PRV1931 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV1931 10 μg
1 options
Catalog No. PRV1931 Supplier Promega Supplier No. V1931
Recombinant full-length human Aurora A was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. Aurora A belongs to a multigenic family of mitotic serine/threonine kinases, which are involved in the control of chromosome segregation.
  • Easily Screen and Profile Aurora A Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg