Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aurora A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517783 Supplier Invitrogen™ Supplier No. PA517783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

AURKA (Aurora A) is a serine/threonine kinase that regulates mitosis by association in vivo with centrosomes. Overexpression of AURKA is observed in many types of cancer. Aurora A plays a role in cell cycle regulation during anaphase and/or telophase, in relation to the function of the centrosome/spindle pole region during chromosome segregation. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal. Aurora A is viewed as a potential target for anticancer drug treatment. Tissue specificity: Highly expressed in testis and weakly in skeletal muscle, thymus and spleen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aurora A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene AURKA
Gene Accession No. O14965
Gene Alias Aik; Airk; AIRK1; Ark1; ARK-1; AU019385; AURA; AURAK; AURKA; Aurora 2; aurora A; Aurora A Kinase; aurora family kinase 1; Aurora kinase A; aurora/IP; aurora/IPL1-like kinase; aurora/IPL1-related kinase 1; AURORA2; Aurora-A; aurora-related kinase 1; AW539821; Ayk1; Breast tumor-amplified kinase; BTAK; EC 2.7.11.1; hARK1; IAK; IAK1; Ipl1- and aurora-related kinase 1; IPL1-related kinase; MGC34538; OTTHUMP00000031341; OTTHUMP00000166072; PPP1R47; protein phosphatase 1, regulatory subunit 47; ratAurA; RP5-1167H4.6; sereine threonine kinase; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; serine/threonine-protein kinase 6; aurora kinase A; serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase Ayk1; STK15; STK6; STK7; Unknown (protein for MGC:134472)
Gene Symbols AURKA
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the amino terminus of human Aurora A/AIK.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6790
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.