Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora A Polyclonal Antibody, HRP, Bioss
SDP

Catalog No. 502281873
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1873 Supplier Bioss Supplier No. BS22695RHRP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aurora A Polyclonal antibody specifically detects Aurora A in Mouse samples. It is validated for Western Blot

Specifications

Antigen Aurora A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene AURKA
Gene Accession No. P97477
Gene Alias Aik; Airk; AIRK1; Ark1; ARK-1; AU019385; AURA; AURAK; AURKA; Aurora 2; aurora A; Aurora A Kinase; aurora family kinase 1; Aurora kinase A; aurora/IP; aurora/IPL1-like kinase; aurora/IPL1-related kinase 1; AURORA2; Aurora-A; aurora-related kinase 1; AW539821; Ayk1; Breast tumor-amplified kinase; BTAK; EC 2.7.11.1; hARK1; IAK; IAK1; Ipl1- and aurora-related kinase 1; IPL1-related kinase; MGC34538; OTTHUMP00000031341; OTTHUMP00000166072; PPP1R47; protein phosphatase 1, regulatory subunit 47; ratAurA; RP5-1167H4.6; sereine threonine kinase; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; serine/threonine-protein kinase 6; aurora kinase A; serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase Ayk1; STK15; STK6; STK7; Unknown (protein for MGC:134472)
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse Aurora A
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20878
Target Species Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.