Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aurora B Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500185JF646
Description
Aurora B Polyclonal antibody specifically detects Aurora B in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Aurora B | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5 | |
The immunogen recognized by this antibody maps to a region between residue 300 and the C-terminus (residue 344) of human Aurora B kinase using the numbering given in Swiss-Prot entry Q96GD4 (GeneID 9212). | |
0.1 mL | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers | |
9212 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction