Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aurora B Monoclonal Antibody (3F11)
Mouse Monoclonal Antibody
$526.00 - $1800.00
Specifications
Antigen | Aurora B |
---|---|
Clone | 3F11 |
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA547316
|
Invitrogen™
MA547316 |
500 μL |
Each for $1,800.00
|
|
|||||
PIMA547414
|
Invitrogen™
MA547414 |
100 μL |
Each for $526.00
|
|
|||||
Description
Aurora B Monoclonal Antibody for Western Blot, ICC/IF, IHC
The Aurora kinases associate with microtubules during chromosome movement and segregation. AURKB (Aurora B) localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers. This complex functions in chromosome alignment and separation, histone modification, and cytokinesis. CPC also includes three non-enzymatic subunits known as the inner centromere protein (INCENP), survivin, and borealin, which determine the activity, localization, stability, and possibly even the substrate specificity of AurB. Two CPCs exist during mitosis: one containing all four members and another consisting of INCENP and AurB. Quaternary CPC functions during chromosome alignment and cytokinesis, whereas the INCENP-AurB complex may be responsible for modifying histone H3. AurB is overexpressed in a number of tumours, including primary breast and colon tumour samples. Its expression in tumours seems to be linked with Aurora-A kinase, suggesting a feedback mechanism between the two proteins.Specifications
Aurora B | |
1 mg/mL | |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 5mM sodium azide | |
O70126, Q96GD4 | |
100073050, 20877, 479492, 9212 | |
Full length recombinant human aurora B protein expressed in and purified from E. coli. | |
Antibody |
3F11 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Monoclonal | |
Liquid | |
IgG2a | |
Human, Mouse, Canine, Horse | |
AURKB | |
Aik2; AIM1; AIM-1; AIRK2; AL022959; ARK2; ARK-2; AurB; AURKB; aurkb-sv1; aurkb-sv2; aurora 1; aurora- and IPL1-like midbody-associated protein 1; aurora B; aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; aurora/IPL1-related kinase 2; aurora-1; aurora-B; aurora-related kinase 2; IPL1; PPP1R48; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase; serine/threonine kinase 12; serine/threonine kinase 5; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 5; serine/threonine-protein kinase aurora-B; STK1; STK-1; Stk12; STK12; AIK2; AIM1; ARK2; IPL1; AIM-1; STK5 | |
AURKB | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title