Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora B Rabbit anti-Human, Polyclonal, FabGennix

Catalog No. 502206171
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6171 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6171 Supplier Fabgennix Inc Supplier No. AURKB201AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aurora B Polyclonal antibody specifically detects Aurora B in Human samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Aurora B
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene AURKB
Gene Accession No. Q96GD4
Gene Alias AIE2; AIK1; Aik2; AIK3; AIM1; AIM-1; AIRK2; AIRK3; AL022959; ARK2; ARK-2; ARK3; ARK-3; AurB; AURKB; aurkb-sv1; aurkb-sv2; AURKC; aurora 1; Aurora 3; aurora- and IPL1-like midbody-associated protein 1; aurora B; aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; Aurora kinase C; aurora/IPL1-related kinase 2; aurora-1; aurora-B; aurora-related kinase 2; EC 2.7.11.1; IPL1; NP_004208; PPP1R48; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase; serine/threonine kinase 12; serine/threonine kinase 5; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 5; serine/threonine-protein kinase aurora-B; STK1; STK-1; Stk12; STK12; AIK2; AIM1; ARK2; IPL1; AIM-1; STK13; STK5
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 300-344 on human Aurora Kinase B protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9212
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.