Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora B Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABS1507 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS1507 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1507 Supplier MilliporeSigma Supplier No. ABS1507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Aurora B is a rabbit polyclonal antibody that detects Aurora Kinase B and has been tested for use in Immunocytochemistry and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aurora B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols AURKB,AIK2,AIM1,AIRK2,ARK2,STK1,STK12,STK5
Host Species Rabbit
Immunogen KLH-conjugated Linear Peptide corresponding to the 11 amino acids from the C-terminal region of Human Aurora kinase B.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001243763.1.
Test Specificity This rabbit polyclonal antibdy detects Aurora kinase B in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.