Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aurora B Recombinant Superclonal™ Antibody (18HCLC)

Catalog No. PI711154
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI711154 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI711154 Supplier Invitrogen™ Supplier No. 711154
Add to Cart
Add to Cart

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Monkey, Horse, Cat and Bovine Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

The Aurora kinases associate with microtubules during chromosome movement and segregation. AURKB (Aurora B) localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers. This complex functions in chromosome alignment and separation, histone modification, and cytokinesis. CPC also includes three non-enzymatic subunits known as the inner centromere protein (INCENP), survivin, and borealin, which determine the activity, localization, stability, and possibly even the substrate specificity of AurB. Two CPCs exist during mitosis: one containing all four members and another consisting of INCENP and AurB. Quaternary CPC functions during chromosome alignment and cytokinesis, whereas the INCENP-AurB complex may be responsible for modifying histone H3. AurB is overexpressed in a number of tumours, including primary breast and colon tumour samples. Its expression in tumours seems to be linked with Aurora-A kinase, suggesting a feedback mechanism between the two proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aurora B
Applications Western Blot
Classification Recombinant Superclonal
Clone 18HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene AURKB
Gene Accession No. O55099, O70126, Q96GD4
Gene Alias Aik2; AIM1; AIM-1; AIRK2; AL022959; ARK2; ARK-2; AurB; AURKB; aurkb-sv1; aurkb-sv2; aurora 1; aurora- and IPL1-like midbody-associated protein 1; aurora B; aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; aurora/IPL1-related kinase 2; aurora-1; aurora-B; aurora-related kinase 2; IPL1; PPP1R48; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase; serine/threonine kinase 12; serine/threonine kinase 5; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 5; serine/threonine-protein kinase aurora-B; STK1; STK-1; Stk12; STK12; AIK2; AIM1; ARK2; IPL1; AIM-1; STK5
Gene Symbols AURKB
Host Species Rabbit
Immunogen Protein corresponding to Human AURKB (aa 89-251).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114592, 20877, 9212
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.