Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aurora C Antibody (OTI2B7), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271303V
Description
Aurora C Monoclonal specifically detects Aurora C in Human samples. It is validated for Western Blot.Specifications
Aurora C | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
AURKC | |
Full-length protein expressed in 293T cell transfected with human Aurora C expression vector. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2B7 | |
Western Blot | |
AIE2AIK3, ARK3Serine/threonine-protein kinase aurora-C, AurC, aurora kinase CAurora/IPL1/Eg2 protein 2, Aurora/IPL1-related kinase 3, aurora-C, Aurora-related kinase 3, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 13 (aurora/IPL1-like), serine/threonine-protein kinase 13, STK13ARK-3 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
6795.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction