Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aurora C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114014 Supplier Invitrogen™ Supplier No. PA5114014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TYDEKVDLWC IGVLCYELLV GYPPFESASH SETYRRILKV DVRFPLSMPL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Goat: 79%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%; Zebrafish: 79%.

AURKC (Aurora C) is a chromosomal passenger protein that forms complexes with AURKB (Aurora B) and inner centromere proteins, playing a role in organizing microtubules in relation to centrosome/spindle function during mitosis. It is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction. Studies on the temporal expression pattern and subcellular localization of Aurora kinases in mitotic cells suggest an association with mitotic structure. Aurora kinase functional influences span from G2 phase to cytokinesis and may be involved in key cell cycle events such as centrosome duplication, chromosome bi-orientation and segregation, cleavage furrow positioning, and ingression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aurora C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene AURKC
Gene Accession No. Q9UQB9
Gene Alias AIE1; AIE2; AIK3; AIRK3; ARK3; ARK-3; AurC; AURKC; Aurora 3; aurora B; aurora kinase C; aurora/Ipl1/Eg2 protein 1; aurora/IPL1/EG2 protein 2; aurora/IPL1-related kinase 3; aurora-C; aurora-related kinase 3; epididymis secretory protein Li 90; HEL-S-90; IAK3; serine/threonine kinase 13 (aurora/IPL1-like); serine/threonine kinase 13 (aurora/IPL-like); serine/threonine-protein kinase 13; Serine/threonine-protein kinase aurora-C; SPGF5; STK13
Gene Symbols AURKC
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human AURKC.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6795
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.