Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aurora C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984939
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4939 Supplier Bioss Supplier No. BS2750R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aurora kinases belong to a highly conserved family of mitotic serine/threonine kinases with three members identified among mammals: Aurora A, B, and C. Studies on the temporal expression pattern and subcellular localization of Aurora kinases in mitotic cells suggest an association with mitotic structure. Aurora kinase functional influences span from G2 phase to cytokinesis and may be involved in key cell cycle events such as centrosome duplication, chromosome bi-orientation and segregation, cleavage furrow positioning, and ingression.

Specifications

Antigen Aurora C
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AURKC
Gene Accession No. Q9UQB9
Gene Alias AIE1; AIE2; AIK2; AIK3; AIM1; AIM-1; AIRK2; AIRK3; ARK2; ARK-2; ARK3; ARK-3; AurC; AURKB; AURKC; Aurora 1; Aurora 3; aurora B; Aurora kinase B; Aurora kinase C; aurora/Ipl1/Eg2 protein 1; Aurora/IPL1/Eg2 protein 2; Aurora/IPL1-related kinase 3; aurora-C; aurora-related kinase 3; epididymis secretory protein Li 90; HEL-S-90; IAK3; serine/threonine kinase 13 (aurora/IPL1-like); serine/threonine kinase 13 (aurora/IPL-like); Serine/threonine-protein kinase 13; Serine/threonine-protein kinase aurora-C; SPGF5; STK1; STK-1; STK12; STK13; STK5
Gene Symbols AURKC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Aurora C .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6795
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.