Learn More
Stoelting™ Autoclip Applier and Accessories
SDP
Autoclip™ Applier works like a staple gun to rapidly apply clip closures.
$335.50 - $665.50
Specifications
Model Autoclip
View More Specs
Products 3
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
10-001-030
SDP
View Documents
Stoelting™
59043
Applier
Each for $665.50
Only null left
Add to Cart
10-001-031
SDP
View Documents
Stoelting™
59046
Remover
Each for $335.50
Only null left
Add to Cart
10-001-032
SDP
View Documents
Stoelting™
59047
Autoclips
Pack of 1000 for $665.50
Only null left
Add to Cart
Description
Description
  • Applier holds 20, 9mm clips, which are held firm and ready for use by rustless beryllium-copper springs
  • Provides maximum holding force with minimal trauma
  • No need for assistance to operate the applier or remover
  • When the wound is healed, Clips are easily removed with the Autoclip or EZ Clip Remover.
Specifications
Specifications
Autoclip
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.