Learn More
Thermo Scientific™ Automation Tips for Molecular Devices™ Pipetting System
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Molecular Devices™ Pipetting System, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Molecular Device FLIPR with Type A CCS 384-well head and Type B Liberty 384-well head.
- Available in non-sterile, sterile and with the Thermo Scientific™ ART™ filter technology
- Low percentage coefficient of variation (%CV)

Specifications
Specifications
Volume (Metric) | 20 μL |
Filtered | Non-filtered |
Sterility | Sterile |
Color | Clear |
Format | Rack |
Tip Style | Automation |
For Use With (Equipment) | Molecular Devices FLIPR Type A CCS 384-well Head |
Product Line | Molecular Devices |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.