Learn More
Axygen™ Automation Tips for PerkinElmer™, 384-Well
GREENER_CHOICE
Catalog No. 10320174
Click to view available options
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Automation Tips for PerkinElmer™, 384-Well are racks of non-filtered, 30μL robotic pipette tips. Constructed of clear polypropyle to stringent specifications. Available in sterile and non-sterile options.
  • Tip format: 96 tip and 384 tip
  • Tip volume range: 20 μL to 235 μL
  • Tip material: Clear or black polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: MDT-96 P50, MDT-384-30 μL
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
Order Info
Rack: 384 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Compliance
Axygen products are manufactured in a clean, controlled state of the art facility and certified RNase-, DNase- and endotoxin-safe
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 30 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Features Standard
Format Rack
Tip Style Standard, Round
Packaging SBS Rack
Quantity 50 Racks/Case
Material Polyethylene
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.