Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Autophinib
GREENER_CHOICESDP

Catalog No. 632450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 63-245-0 Supplier Tocris Bioscience™ Supplier No. 6324/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent VPS34 inhibitor

Autophinib is a potent VPS34 inhibitor (IC50 = 19 nM). Inhibits starvation- and Rapamycin -induced autophagy (IC50 values = 40 and 90 nM, respectively).

Specifications

Chemical Name or Material 6-Chloro-N-(5-methyl-1H-pyrazol-3-yl)-2-(4-nitrophenoxy)-pyrimidinamine
CAS 1644443-47-9
Quantity 50 mg
Molecular Formula C14H11ClN6O3
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.