Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ AutoRED AP Plus Staining Kit (Colorimetric)
SDP

Catalog No. NB397844 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397844 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397844 Supplier Novus Biologicals™ Supplier No. NBP312172
Add to Cart
Add to Cart

Assay Kit (Colorimetric)

Specifications

Content And Storage Store at 4°C.
For Use With (Application) Immunohistochemistry (IHC), Immunohistochemistry (Paraffin), ISH
Quantity 30 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.