Learn More
Trajan Scientific and Medical™ Autosampler Syringes for Agilent GCs
SDP
Catalog No. 03350408 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
Needle Gauge:
23 G
26 G
Needle Point Style:
Cone
Side Hole
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Needle Gauge Needle Point Style
03-350-408 10 μL 23 G Side Hole
03-350-400 5 μL 26 G Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-350-408 Supplier Trajan Scientific and Medical™ Supplier No. 002811
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes for Agilent GCs has the knowledge and expertise to manufacture an SGE syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Agilent instruments
Specifications
Needle Gauge 23 G
Diameter (Metric) Outer Needle 0.63 mm
Product Type Fixed Needle Syringe
Volume (Metric) 10 μL
Length (Metric) Needle 42 mm
Needle Point Style Side Hole