Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Cone Tipped Needle for Agilent GCs
SDP
Catalog No. 03350407 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
10 μL
Catalog No. Volume (Metric)
03-350-407 10 μL
1 options
Catalog No. 03-350-407 Supplier Trajan Scientific and Medical™ Supplier No. 002820
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Cone Tipped Needle for Agilent GCs has the knowledge and expertise to manufacture an SGE syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Agilent instruments
Specifications
Needle Gauge 23 G
Diameter (Metric) Inner Needle 0.11 mm
Diameter (Metric) Outer Needle 0.63 mm
Product Type Removable Needle Agilent Syringe with GT Plunger
Volume (Metric) 10 μL
Needle Point Style Cone