Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Autotaxin (ENPP2) Mouse anti-Mouse, Human, Unconjugated, Clone: 7A5, MilliporeSigma™

Catalog No. MABT1350100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT1350-100 100 μg
MABT1350-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT1350-100 Supplier MilliporeSigma Supplier No. MABT1350100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Autotaxin (ENPP2), clone 7A5, Cat. No. MABT1350, is a mouse monoclonal antibody that detects Autotaxin (Ectonucleotide pyrophosphatase/phosphodiesterase family member 2) and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Autotaxin (ENPP2)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7A5
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Ectonucleotide pyrophosphatase/phosphodiesterase family member 2;EC: 3.1.4.39;E-NPP 2;Extracellular lysophospholipase D;LysoPLD
Gene Symbols ENPP2;ATX;PDNP2
Host Species Mouse
Immunogen His-tagged recombinant fragment (without the first 28 amino acids) from human Autotaxin.
Molecular Weight of Antigen ∽100 kDa observed; 98.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001035181
Test Specificity Clone 7A5 detects human and murine autotaxin.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.