Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AUTS2 Antibody (CL7067), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | AUTS2 |
---|---|
Clone | CL7067 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Host Species | Mouse |
Description
AUTS2 Monoclonal specifically detects AUTS2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
AUTS2 | |
Monoclonal | |
Mouse | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
26053 | |
This antibody was developed against a synthetic peptide corresponding to amino acids: GISPLPGGERFPYPSFHWD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL7067 | |
Unconjugated | |
Human, Mouse | |
autism susceptibility candidate 2, autism susceptibility gene 2 protein, autism-related protein 1, KIAA0442FBRSL2, MGC13140 | |
AUTS2 | |
IgG1 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title