Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Auxin response factor Monoclonal Antibody (14D1)
GREENER_CHOICE

Catalog No. PIMA534936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534936 Supplier Invitrogen™ Supplier No. MA534936
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: Tea leafbud.

Auxin response factors (ARFs) are transcription factors that bind to auxin response elements in promoters of early auxin response genes and play central roles in many auxin-mediated processes. As transcription factors, ARFs possess a modular structure and consist of three major domains: an amino-terminal DNA-binding domain (DBD); a middle domain, which is responsible for gene activation or repression; and a carboxy-terminal PB1(Phox and Bem1) domain, contained within a region that was previously called domain III/I. ARFs function as transcription activators or repressors, depending on the middle region of the ARF protein; Q-rich middle regions activate transcription, whereas P/S/T-rich middle regions confer repressor activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Auxin response factor
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 14D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Host Species Mouse
Immunogen Recombinant protein within tea Auxin response factor aa 401-586.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.