Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Auxin
GREENER_CHOICESDP

Catalog No. 683450 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-345-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 68-345-0 Supplier R&D Systems™ Supplier No. 6834/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Chemical dimerizer used in auxin-inducible degron (AID) system; phytohormone

Auxin is a chemical dimerizer used in auxin-inducible degron (AID) systems. Induces degradation of a target protein tagged with the auxin-receptor F-box protein Tir1 E3 ligase AID in human colorectral cancer and mouse ES cells or tagged with AFB2 in A431 cells. Endogenous plant hormone.

Specifications

Chemical Name or Material 1 H-Indole-3-acetic acid
CAS 87-51-4
Quantity 50 mg
Product Line Tocris
Target Protein Dimerizers
Molecular Formula C10H9NO2
Formula Weight 175.18
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.