Learn More
Heidolph™ AVA Software Kit - US Plug
SDP
Heidolph™ AVA Software Kit - US Plug includes Controller, Data Hub, and AVA Installed
Supplier:  Heidolph™ 015200006
 View more versions of this product
Catalog No. 13-876-719
Only null left
Add to Cart
Request a Quote
Description
Description
  • AVA easily automates chemistry procedures
  • Makes chemistry safer by running unattended experiments safely, reducing manual errors, setting apparatus safety limits to prevent accidents
  • Configures audio and visual alarms
  • Advantages of software automation include improved productivity, and easy results sharing between users which improves research/collaboration
  • AVA software can:
    • Control, log common laboratory apparatus and processes
    • Eliminate time-consuming, manual data recording
    • Track what you do, as you do it and repeat in the experiment scheduler
    • Visualize results for rapid analysis and reporting
    • Link devices for dual control (pump and balance for additions)
    • Control up to four experiments in parallel
Specifications
Specifications
Level 2
Controller, Data Hub, and AVA Installed
AVA
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.