Learn More
Heidolph™ AVA Software Kit - US Plug
SDP
Heidolph™ AVA Software Kit - US Plug includes Controller, Data Hub, and AVA Installed
$6675.43 - $20846.17
Specifications
Includes Controller, Data Hub, and AVA Installed
Product Line AVA
View More Specs
Products 4
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
13-876-718
SDP
View Documents
Heidolph™
015200005
Level 1
Each for $6,675.43
Only null left
Add to Cart
13-876-719
SDP
View Documents
Heidolph™
015200006
Level 2
Each for $7,914.52
Only null left
Add to Cart
13-876-720
SDP
View Documents
Heidolph™
015200007
Level 3
Each for $13,418.84
Only null left
Add to Cart
13-876-721
SDP
View Documents
Heidolph™
015200008
Level 4
Each for $20,846.17
Only null left
Add to Cart
Description
Description
  • AVA easily automates chemistry procedures
  • Makes chemistry safer by running unattended experiments safely, reducing manual errors, setting apparatus safety limits to prevent accidents
  • Configures audio and visual alarms
  • Advantages of software automation include improved productivity, and easy results sharing between users which improves research/collaboration
  • AVA software can:
    • Control, log common laboratory apparatus and processes
    • Eliminate time-consuming, manual data recording
    • Track what you do, as you do it and repeat in the experiment scheduler
    • Visualize results for rapid analysis and reporting
    • Link devices for dual control (pump and balance for additions)
    • Control up to four experiments in parallel
Specifications
Specifications
Controller, Data Hub, and AVA Installed
AVA
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.