Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AVEN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176764
Description
AVEN Polyclonal specifically detects AVEN in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
AVEN | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
apoptosis, caspase activation inhibitor, cell death regulator Aven, PDCD12, programmed cell death 12 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
NP_065104 | |
AVEN | |
Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human Aven. The immunogen is located within the first 50 amino acids of Aven. Amino Acid Squence: QAERGARGGRGRRP | |
0.1 mg | |
Apoptosis | |
57099 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction