Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AVEN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582735
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582735 Supplier Invitrogen™ Supplier No. PA582735
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPSRDSQKPT SPLQSAGDHL EEELDLLLNL DAPIKEGDNI LPDQTSQDLK SKEDGEVVQE EEVCAKPSVT EEKNMEPEQP STSK Highest antigen sequence indentity to the following orthologs: Mouse - 62%, Rat - 63%.

Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Aven is a recently discovered protein that blocks apoptosis induced by Apaf-1 and caspase-9. It is thought that Aven functions by binding to Bcl-xL, an antiapoptotic member of the Bcl-2 family, and to Apaf-1, possibly interfering with the ability of Apaf-1 to self-associate, suggesting that Aven impedes Apaf-1-mediated caspase activation. Higher levels of Aven mRNA are seen in patients with acute leukemia than in control patients, suggesting that Aven may be useful as a prognostic indicator in leukemia patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AVEN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AVEN
Gene Accession No. Q9NQS1
Gene Alias 1700013A01Rik; 1700056A21Rik; apoptosis and caspase activation inhibitor; apoptosis, caspase activation inhibitor; AVEN; Cell death regulator Aven; mAven-L; mAven-S; PDCD12; programmed cell death 12
Gene Symbols AVEN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AVEN. Recombinant protein control fragment (Product #RP-92081).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57099
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.