Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Avermectin Monoclonal Antibody (1G8)
GREENER_CHOICE

Catalog No. PIMA549736
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549736 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549736 Supplier Invitrogen™ Supplier No. MA549736
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Avermectin is a group of macrocyclic lactone compounds that are used as broad-spectrum anthelmintics, insecticides, and acaricides. These compounds are derived from the soil bacterium Streptomyces avermitilis and are commonly used in veterinary medicine to treat parasitic infections in livestock and pets. Avermectin compounds work by binding to glutamate-gated chloride ion channels in the nervous system of parasites, causing paralysis and death. Avermectin compounds have also been found to have potential as a treatment for certain human diseases, such as river blindness and lymphatic filariasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Avermectin
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 1G8
Concentration 2.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of AVM hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.