Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avi Epitope Tag Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326066AF594
Description
Avi Epitope Tag Polyclonal antibody specifically detects Avi Epitope Tag in Epitope Tag samples. It is validated for Western BlotSpecifications
Avi Epitope Tag | |
Polyclonal | |
Western Blot | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Tag | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Produced in rabbits immunized with a synthetic peptide corresponding to the Avi Epitope Tag sequence (GLNDIFEAQKIEWHE) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction