Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avi Epitope Tag Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$110.20 - $565.00
Specifications
Antigen | Avi Epitope Tag |
---|---|
Dilution | Western Blot 1:1000-1:5000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB293793
![]() |
Novus Biologicals
NBP326066500UL |
500 μL |
Each for $565.00
|
|
|||||
NB293795
![]() |
Novus Biologicals
NBP326066100UL |
100 μL |
Each for $322.50
|
|
|||||
NB293794
![]() |
Novus Biologicals
NBP32606620UL |
20 μL |
Each for $110.20
|
|
|||||
Description
Avi Epitope Tag Polyclonal antibody specifically detects Avi Epitope Tag in Epitope Tag samples. It is validated for Western BlotSpecifications
Avi Epitope Tag | |
Western Blot | |
Unconjugated | |
Rabbit | |
Tag | |
Produced in rabbits immunized with a synthetic peptide corresponding to the Avi Epitope Tag sequence (GLNDIFEAQKIEWHE) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 1:1000-1:5000 | |
Polyclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title