Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avi tag Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5118383

Description
This product is predicted to work in ELISA at a dilution of 1:10,000. The amino acid sequence for this product is: GLNDIFEAQKIEWHE.
The Avi tag is a biotin-acceptor peptide1, GLNDIFEAQKIEWHE. The 15-residue peptide served as a substrate mimic for biotin ligase (BirA), which usually recognizes the much larger protein domain.
Specifications
Avi tag | |
Polyclonal | |
Unconjugated | |
Avi; BirA | |
This antibody is produced by immunizing rabbits with a KLH-coupled synthetic peptide | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Tag | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction