Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241050
Description
Avian Influenza Hemagglutinin Polyclonal specifically detects Avian Influenza Hemagglutinin in Virus samples. It is validated for ELISA.Specifications
Avian Influenza Hemagglutinin | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Antibody was raised against a synthetic peptide corresponding to 14 amino acids in the middle of the avian influenza hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 320 - 370 of Avian Influenza Hemagglutinin. Amino Acid Squence: RNSPQRERRRKKRG | |
0.1 mg | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
ELISA | |
1 mg/mL | |
ELISA 1 μg/mL | |
Rabbit | |
Affinity Purified | |
RUO | |
Virus | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction