Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP2410660.025MG
Description
Avian Influenza Hemagglutinin 4 Polyclonal specifically detects Avian Influenza Hemagglutinin 4 in Virus samples. It is validated for Western Blot, ELISA.Specifications
| Avian Influenza Hemagglutinin 4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against a synthetic peptide corresponding to 10 amino acids near the center of the Hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 260 - 310 of Avian Influenza Hemagglutinin 4. Amino Acid Squence: REDIMKSELEYGN | |
| 0.025 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/mL | |
| Western Blot, ELISA 1 μg/mL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction