Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Avian Influenza Hemagglutinin 1 |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Avian Influenza Hemagglutinin 1 Polyclonal specifically detects Avian Influenza Hemagglutinin 1 in Virus samples. It is validated for ELISA.Specifications
Avian Influenza Hemagglutinin 1 | |
ELISA | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
IgG | |
Affinity Purified |
1 mg/mL | |
Polyclonal | |
Goat | |
Virus | |
Antibody was raised against a synthetic peptide corresponding to 14 amino acids in the middle of the Avian Influenza Hemagglutinin 1 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 320 - 370 of Avian Influenza Hemagglutinin 1. Amino Acid Squence: RNSPQRERRRKKRG | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title