Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza A H5N1 Hemagglutinin Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241050MFV450
Description
Avian Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Avian Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for ELISASpecifications
Avian Influenza A H5N1 Hemagglutinin | |
Polyclonal | |
ELISA | |
Harvey rat sarcoma viral oncogene homolog | |
Antibody was raised against a synthetic peptide corresponding to 14 amino acids in the middle of the avian influenza hemagglutinin protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 320 - 370 of Avian Influenza Hemagglutinin. Amino Acid Squence: RNSPQRERRRKKRG | |
0.1 mL | |
Infections (Virus Bacteria and Parasites) | |
Virus | |
Purified |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction