Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza A H5N1 M2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241074MFV450
Description
Avian Influenza A H5N1 M2 Polyclonal antibody specifically detects Avian Influenza A H5N1 M2 in Virus samples. It is validated for ELISASpecifications
| Avian Influenza A H5N1 M2 | |
| Polyclonal | |
| ELISA | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Antibody was raised against a 13 amino acid synthetic peptide from amino terminus of H5N1 M2. The immunogen is located within the first 50 amino acids of Avian Influenza A M2. Amino Acid Squence: LTEVETPTRNEWE | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction