Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza A H5N1 Neuraminidase Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241048MFV450
Description
Avian Influenza A H5N1 Neuraminidase Polyclonal antibody specifically detects Avian Influenza A H5N1 Neuraminidase in Virus samples. It is validated for ELISASpecifications
Avian Influenza A H5N1 Neuraminidase | |
Polyclonal | |
ELISA | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Antibody was raised against a synthetic peptide corresponding to 16 amino acids in the middle of the avian influenza neuraminidase protein. Efforts were made to use relatively conserved regions as the antigen. The immunogen is located within amino acids 150 - 200 of Avian Influenza Neuraminidase. Amino Acid Squence: CPIGESPSPYNSRFES | |
0.1 mL | |
Infections (Virus Bacteria and Parasites) | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction