Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avian Influenza A H6N1 Nucleoprotein Antibody, Novus Biologicals™
SDP

Catalog No. NB10056611 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056611 0.1 mg
NB10056611T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056611 Supplier Novus Biologicals Supplier No. NB10056611
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Avian Influenza A H6N1 Nucleoprotein Polyclonal antibody specifically detects Avian Influenza A H6N1 Nucleoprotein in Virus samples. It is validated for Western Blot.

Specifications

Antigen Avian Influenza A H6N1 Nucleoprotein
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Alias NP, Nucleoprotein
Host Species Rabbit
Immunogen Amino acids 58-77 of avian flu nucleoprotein (accession number Q71FW9) were used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is 95% homologous to chicken and quail H5N1 and 90% homologous to swine H5N1.
Target Species Virus, Chicken
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.