Learn More
Novus Biologicals™ Avian Influenza A H7N9 Hemagglutinin - (A/Anhui/1/2013) ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 15.6-1000 pg/mL |
| Assay Sensitivity | 4.9 pg/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Recombinant Protein |
| Specificity | Recognizes both recombinant and natural Avian Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin. |
| For Use With (Application) | ELISA, Sandwich ELISA |
| Quantity | 1 Kit |
| Research Discipline | Infections (Virus Bacteria and Parasites) |
| Storage Requirements | Storage is content dependent. |
| Synonym | H7N9HA |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.