Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza A H7N9 Hemagglutinin Antibody (001)

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289789
Description
Avian Influenza A H7N9 Hemagglutinin Monoclonal specifically detects Avian Influenza A H7N9 Hemagglutinin in Virus samples. It is validated for ELISA.Specifications
Avian Influenza A H7N9 Hemagglutinin | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Has cross-reactivity in ELISA with: H7N9 (A/Anhui/1/2013) HA H7N9 (A/Shanghai/1/2013) HA H7N7 (A/chicken/Netherlands/1/03) HA H7N7 (A/Netherlands/219/03) HA H10N3 (A/duck/Hong Kong/786/1979) HA H15N8 (A/duck/AUS/341/1983) HA No cross-reactivity in ELISA with: H1N1 (A/California/07/2009) HA H2N2 (A/Canada/720/2005) HA H3N2 (A/Brisbane/10/2007) HA H4N6 (A/mallard/Ohio/657/2002) HA H5N1 (A/Anhui/1/2005) HA H6N1 (A/northern shoveler/California/HKWF115/2007) HA H8N4 (A/pintail duck/Alberta/114/1979) HA H9N2 (A/Hong Kong/1073/99) HA H11N2 (A/duck/Yangzhou/906/2002) HA H12N5 (A/green-winged teal/ALB/199/1991) HA H13N8 (A/black-headed gull/Netherlands/1/00) HA H16N3 (A/black-headed gull/Sweden/5/99) HA | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1 | |
ELISA 1:5000-1:10000 | |
H7N9HA | |
Recombinant H7N9 Avian Influenza A H7N9 Hemagglutinin | |
50 μL | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction