Learn More
Invitrogen™ Avian Influenza H5N1 HA Monoclonal Antibody (14B1E2G6)

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for H5N1 (A/Anhui/1/2005) HA.
Specifications
Specifications
| Antigen | Avian Influenza H5N1 HA |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 14B1E2G6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | Avian H5N1; flu hemagglutinin; H5N1; influenza a; Influenza A Neuraminidase; influenza hemagglutinin |
| Host Species | Mouse |
| Immunogen | Recombinant H5N1 HA protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.