Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza NS1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241067MFV500
Description
Avian Influenza NS1 Polyclonal antibody specifically detects Avian Influenza NS1 in Virus samples. It is validated for ELISASpecifications
Avian Influenza NS1 | |
Polyclonal | |
ELISA | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Virus | |
Purified |
ELISA | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Antibody was raised against a synthetic peptide corresponding to 14 amino acids at the carboxy terminus of the Avian Influenza Nonstructural Protein 1 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within the last 50 amino acids of Avian Influenza Nonstructural Protein 1. Amino Acid Squence: LPPNQKRKMARTIE | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction