Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Avian Influenza NS2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241068MFV450
Description
Avian Influenza NS2 Polyclonal antibody specifically detects Avian Influenza NS2 in Virus samples. It is validated for ELISASpecifications
Avian Influenza NS2 | |
Polyclonal | |
ELISA | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Virus | |
Purified |
ELISA | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Antibody was raised against a synthetic peptide corresponding to 14 amino acids near the middle of the Avian Influenza Nonstructural Protein 2 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within amino acids 40 - 90 of Avian Influenza Nonstructural Protein 2. Amino Acid Squence: RNGKWREQLSQKFE | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction