Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Checkers Aviation Wheel Chocks for Mid/Large-Sized Aircraft AC4600 Checkers
GREENER_CHOICE

Catalog No. 19088483 Shop All Justrite Safety Group Products
Click to view available options
Color:
Yellow

Aviation Wheel Chocks include the AC series, made of highly durable, lightweight polyurethane.

  • Checkers™ Aviation Wheel Chocks include the AC series, made of highly durable, lightweight polyurethane
  • AC Chocks last longer than wood chocks, never need painting and eliminate Foreign Object Damage threats for aircraft
  • The AC Series is ideal for private, commercial, cargo and military aircraft

Specifications

Color Yellow
Product Type Wheel Chock
Dimensions (L x W x H) 14.5 x 6 x 4 in.
Material Polyurethane
For Use With (Equipment) Mid/Large-Sized Aircraft
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.