Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Avidin, FITC conjugate

Catalog No. p-7075642
Encompass_Preferred
Change view
Click to view available options
Packaging:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
A821 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A821 Supplier Invitrogen™ Supplier No. A821
Only null left
Add to Cart
Edge
Add to Cart

Avidin, FITC conjugate

Our avidin, fluorescein conjugate has excitation and emission maxima ∼494/518. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC (Fluorescein)
Quantity 5 mg
Packaging 1 Each
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Type Avidin

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.