Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ eBioscience™ Avidin HRP
Click to view available options
Quantity:
500 μL
1 mL
Description
Avidin binds to biotin with high affinity. Because sodium azide is an inhibitor of horseradish peroxidase enzymatic activity, it should not be added to the AV-HRP diluent.
Specifications
Specifications
| Quantity | 500 μL |
| Target Molecule | Biotin |
| Form | Liquid |
| Product Line | eBioscience™ |
| Label or Dye | HRP (Horseradish Peroxidase) |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction