Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AVIL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563621
Description
Immunogen sequence: HASQDELAAS AYQAVEVDRQ FDGAAVQVRV RMGTEPRHFM AIFKGKLVIF EGGTSRKGNA EPDPPVRLFQ IHGNDKSNTK AVEVPA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.
AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.
Specifications
AVIL | |
Polyclonal | |
Unconjugated | |
AVIL | |
actin-binding protein DOC6; Advil; Advillin; Advillin-like protein; Avil; DOC6; p92; peripheral nervous system villin-like protein; pervin; Unknown (protein for MGC:136857); zgc:136857 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10677 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75366 | |
AVIL | |
Recombinant protein corresponding to Human AVIL. Recombinant protein control fragment (Product #RP-97750). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction