Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AVIL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590703
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590703 Supplier Invitrogen™ Supplier No. PA590703
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HT-29, Mouse uterus, Mouse brain, Rat uterus, Rat testis, Rat brain; Cellular Location: Cell projection, Cytoplasm, axon, cytoskeleton.

AVIL is a member of the gelsolin/villin family of actin regulatory proteins. This protein has structural similarity to villin. It binds actin and may play a role in the development of neuronal cells that form ganglia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AVIL
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AVIL
Gene Accession No. O75366, O88398, Q9WU06
Gene Alias actin-binding protein DOC6; Advil; Advillin; Advillin-like protein; Avil; DOC6; p92; peripheral nervous system villin-like protein; pervin; Unknown (protein for MGC:136857); zgc:136857
Gene Symbols AVIL
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10677, 11567, 79253
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.